missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35764-100ul
This item is not returnable.
View return policy
Description
TOM1L1 Polyclonal antibody specifically detects TOM1L1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| TOM1L1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| OK/KNS-CL.3, Src-activating and signaling molecule protein, SRCASMSrc activating and signaling molecule, target of myb 1-like 1, target of myb1 (chicken) homolog-like 1, target of myb1 (chicken)-like 1, target of myb1-like 1, Target of Myb-like protein 1, TOM1-like protein 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 250-350 of human TOM1L1 (NP_005477.2).,, Sequence:, QERIMDLLVVVENEDVTVELIQVNEDLNNAILGYERFTRNQQRILEQNKNQKEATNTTSEPSAPSQDLLDLSPSPRMPRATLGELNTMNNQLSGLNFSLPS | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10040 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction