Learn More
TOM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17506-1-AP-150UL
 View more versions of this product
Product Code. 16867404
  • £349.00 / 150µL
Description
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
TOM1
Polyclonal
Unconjugated
TOM1
Target of Myb protein 1, target of myb1 (chicken), TOM1
Rabbit
Antigen Affinity Chromatography
RUO
10043, 21968, 361370
-20°C
Liquid
Western Blot
0.17 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O60784, O88746
Tom1
TOM1 Fusion Protein Ag11706
150 μL
Primary
Human, Mouse, Rat