missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TOM1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25813-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| TOM1 | |
| Polyclonal | |
| Unconjugated | |
| TOM1 | |
| Target of Myb protein 1, TOM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10043, 21968 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O60784, O88746 | |
| TOM1 | |
| TOM1 Fusion Protein Ag22931 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction