missing translation for 'onlineSavingsMsg'
Learn More

Tolycaine, MedChemExpress

Product Code. 16470397
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16470397 50 mg 50mg
16424048 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16470397 Supplier Medchem Express Supplier No. HY105584
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tolycaine is an analog of Lidocaine (HY-B0185) and causes convulsions in vivo.

Tolycaine is an analog of Lidocaine (HY-B0185) and causes convulsions in vivo.

Specifications

Chemical Name or Material Tolycaine
CAS 3686-58-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H22N2O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (359.26 mM)
SMILES CCN(CC)CC(NC1=C(C=CC=C1C)C(OC)=O)=O
Molecular Weight (g/mol) 278.35
Formula Weight 278.35
Percent Purity 99.13%
Grade Research
Physical Form Liquid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.