missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Tolvaptan

Description
Tolvaptan is a potent and selective competitive vasopressin V2 receptor antagonist (Ki values are 0.06 and 12.3 nM for V2 and V1a receptors respectively). Decreases urine osmolality and increases serum sodium concentrations. Delays the onset of end-stage renal disease in a mouse model of polycystic kidney disease. Exhibits myocardial and renal protective effects in hypertensive heart failure rats. Orally active.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Tolvaptan |
| Purity | >98% |
| Molecular Weight (g/mol) | 448.94 |
| Inhibitors | Tolvaptan |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?