missing translation for 'onlineSavingsMsg'
Learn More

Toluidine Blue, Pure, C.I.52040, Fisher Chemical

Product Code. 10606662 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10606662 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10606662 Supplier Fisher Chemical Supplier No. T/2660/46
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

25g, Glass Jar

TRUSTED_SUSTAINABILITY

Specifications

CAS 92-31-9
Molecular Formula C15H16ClN3S
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
Molecular Weight (g/mol) 305.82
PubChem CID 7083
Formula Weight 305.82g/mol
Packaging Glass Jar
Color Green
Quantity 25 g
Physical Form Solid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.