missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toluidine Blue O, pure

Product Code. 10613732 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10613732 5 g Glass bottle 5g
10506271 25 g Glass bottle 25g
10135113 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10613732 Supplier Thermo Scientific Chemicals Supplier No. 348600050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-31-9
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
Synonym Basic blue 17, C.I. 52040, Tolonium chloride
PubChem CID 7083
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Beilstein 27, 402
Merck Index 15, 9679
Solubility Information Solubility in water: 38.2g/L. Other solubilities: soluble in alcohol
Infrared Spectrum Authentic
IUPAC Name 3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
Formula Weight 305.82
Grade Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Green
Quantity 5 g
Physical Form Powder
Chemical Name or Material Toluidine blue O
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.