missing translation for 'onlineSavingsMsg'
Learn More

Toluidine Blue O, Fisher BioReagents™

Product Code. 10573355
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10573355 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10573355 Supplier Fisher BioReagents Supplier No. BP10710
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This nuclear stain may be used as an alternative to methylene blue

TRUSTED_SUSTAINABILITY

Specifications

CAS 92-31-9
Extinction Coefficient 700g(-1) cm(-1) min.
Molecular Formula C15H16ClN3S
MDL Number MFCD00011934
Merck Index 15, 9679
Solubility Information Soluble in water
Identification Pass Test
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
Molecular Weight (g/mol) 305.82
PubChem CID 7083
Lambda Max 620 to 638nm
Formula Weight 305.65
Loss on Drying 10% max.
Packaging Glass Bottle
Vapor Pressure Vapor Pressure: Negligible
Color Index No. 52040
Color Green
Quantity 10 g
Physical Form Solid
Chemical Name or Material Toluidine Blue O
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.