missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Toluidine Blue O, Certified, 65% min dye content

Product Code. 15437209
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
Unit Size:
250g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15437209 5 g 5g
15447209 25 g 25g
15457209 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15437209 Supplier Thermo Scientific Chemicals Supplier No. J66015.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92-31-9 | C15H16ClN3S | 305.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-31-9
Molecular Formula C15H16ClN3S
Molecular Weight (g/mol) 305.82
MDL Number MFCD00011934
InChI Key HNONEKILPDHFOL-UHFFFAOYSA-M
Synonym Basic Blue 17,C.I. 52040
PubChem CID 7083
IUPAC Name (7-amino-8-methylphenothiazin-3-ylidene)-dimethylazanium;chloride
SMILES [Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

Specifications

Solubility Information Soluble in water at 1mg/ml
Formula Weight 305.83
Percent Purity ≥65% dye content
Packaging Glass bottle
Quantity 5 g
Physical Form Powder
Chemical Name or Material Toluidine Blue O

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.