missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl mercaptan, 99%, Thermo Scientific Chemicals
32.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Melting Point | -29°C |
Color | Colorless to Yellow |
Density | 1.0580g/mL |
Boiling Point | 195°C |
Flash Point | 70°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Show More |
Safety and Handling
Product Identifier
- Benzyl mercaptan
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P264-Wash thoroughly after handling.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'