missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Spectrophotometric Grade, 99.7+%

Product Code. 11383537
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
4 x 4 L
Unit Size:
1 L
16 L
4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11383537 4 L 4 L
11373537 1 L 1 L
11353537 4 x 1 L 4 L
11363537 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11383537 Supplier Thermo Scientific Alfa Aesar Supplier No. 019376.K7
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03% | CAS: 108-88-3 | C7H8 | 92.14 g/mol

Toluene is used in spectrophotometry and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Tekniske data

Melting Point °C to 93°C
Color Colorless
Appearance Colorless liquid
Density 0.867
Quantity 4 L
Flash Point 4°C (40°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range >99.7%
Beilstein 635760
Boiling Point 111°C
Solubility Information Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Grade Spectrophotometry
Chemical Name or Material Toluene
Vis mere Vis mindre

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.