Product Code. 15621640
Click to view available options
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
20 L
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
15621640
6 x 1 L
Glass Bottle
Pack of 6
15601640
20 L
Stainless Steel Can
20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
15621640
Supplier
Fluka
Supplier No.
1799656X1L
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Laboratory Reagent,≥99.3%
≥95% for general laboratory use.
Chemical Identifiers
CAS
108-88-3
Molecular Formula
C7H8
Molecular Weight (g/mol)
92.14
MDL Number
MFCD00008512
InChI Key
YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID
1140
ChEBI
CHEBI:17578
IUPAC Name
toluene
SMILES
CC1=CC=CC=C1
Specifications
Melting Point
-95°C
Color
Colorless
Density
0.865g/mL at 25°C
Quantity
6 x 1 L
Explosion Limit
0.07
Flash Point
4°C (39.2°F)
Ignition Point
997°F
Packaging
Glass Bottle
Physical Form
Liquid
Refractive Index
n/D 1.496(lit.)
Linear Formula
C6 H5 CH3
Vapor Pressure
22mmHg (20°C), 26mmHg (25°C)
UN Number
UN1294
Beilstein
635760
Boiling Point
110.6°C
Formula Weight
92.14g/mol
Percent Purity
≥99.3%
Grade
Laboratory Reagent
Chemical Name or Material
Toluene
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?