missing translation for 'onlineSavingsMsg'
Learn More

Toluene, Environmental Grade, 99.8+%

Product Code. 11394708
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
Unit Size:
1 L
16 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394708 4 x 4 L 16 L
11384708 1 L 1 L
11304718 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11394708 Supplier Thermo Scientific Alfa Aesar Supplier No. 043061.M6
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Suitable For Pesticide Residue Analysis | CAS: 108-88-3 | C7H8 | 92.14 g/mol

Suitable for pesticide residue analysisToluene is used as a solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as a standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and as a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Maximum level of impurities: Color (APHA) 10, Halogenated residue by GC-ECD, (as Heptachlorepoxide) 5ng/L, Water 0.03%
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point °C to 93°C
Color Colorless
Density 0.867 g/cm3 at 20°C
Quantity 4 x 4 L
Flash Point 4°C (40°F)
Ignition Point 535°C (995°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.4967
Assay Percent Range >99.8%
Vapor Pressure 22mm Hg at 20°C
Beilstein 635760
Boiling Point 111°C
Solubility Information Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Grade Environmental
Chemical Name or Material Toluene, Submicron filtered
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.