missing translation for 'onlineSavingsMsg'
Learn More

Toluene Diisocyanate, TRC

Product Code. 30010867
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010867 10 g 10g
30004766 5 g 5g
30007378 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010867 Supplier Toronto Research Chemicals Supplier No. TRCB42669310G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26471-62-5
Molecular Formula 2 C9 H6 N2 O2
Molecular Weight (g/mol) 348.312
Synonym Diisocyanatotoluene, Methylphenylene ester Isocyanic Acid, Methyl-m-phenylene isocyanate, Methylphenylene isocyanate, Toluene Diisocyanate
IUPAC Name 1,3-diisocyanato-2-methylbenzene;2,4-diisocyanato-1-methylbenzene
SMILES Cc1ccc(cc1N=C=O)N=C=O.Cc2c(cccc2N=C=O)N=C=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/2C9H6N2O2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13;1-7-8(10-5-12)3-2-4-9(7)11-6-13/h2*2-4H,1H3
Formula Weight 348.086
Chemical Name or Material Toluene Diisocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.