missing translation for 'onlineSavingsMsg'
Learn More

Toluene, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. p-7138051 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10170571

Brand: JT Baker 9336.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Toluene, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87g/cm3 at 20°C
Boiling Point 110°C to 111°C
Quantity 1 L
Explosion Limit 1.2 - 7 Vol %
Flash Point 4°C
Ignition Point 535°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Linear Formula C6H5CH3
Viscosity 0.6 mPas at 20°C
Water Max. 0.03%
Solubility Information 0.5g/L in water
Formula Weight 92.14g/mol
Percent Purity ≥99.7%
Grade For Organic Residue Analysis
Assay By GC, corrected for water
Chemical Name or Material Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.