missing translation for 'onlineSavingsMsg'
Learn More

Toluene, BAKER ULTRA RESI-ANALYZED, J.T. Baker™

Product Code. 15548584 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15548584 2.5 L 2.5 L
10170571 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15548584 Supplier JT Baker Supplier No. 9336.2500
  • ONLINE EXCLUSIVE PRICE
    £265.00
    £225.67 / 2.5 L
    Save £39.33
    14% Off
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Gives superior performance with minimal risk of contaminants that can limit accuracy and productivity. Toluene, BAKER ULTRA RESI-ANALYZED, J.T. Baker™ is an ultra-pure solvent for organic residue analysis that maximizes an instrument’s sensitivity and detection power.

BAKER ULTRA RESI-ANALYZED solvents were developed for organic residue extraction/concentration procedures. These solvents are fully characterized and lot controlled by ECD, FID, or other method-specific detectors to deliver the highest level of purity and lot-to-lot consistency.

  • Unique stabilizer systems provide unmatched product stability and interference-free results when chemical requires
  • Optimized distillation targeted to remove reactive impurities, production of higher assays and narrow solvent fronts
  • Performance tested to purity levels below the Lower Limits of Quantitation (LLQ) for trace analyte detection
  • Performance is guaranteed through the expiration period stated on the label
  • Suitable for exacting EPA sample cleanup protocols

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Color Colorless
Density 0.87g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 1.2 - 7 Vol %
Flash Point 4°C
Ignition Point 535°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1 ppm
Linear Formula C6H5CH3
Viscosity 0.6 mPas at 20°C
Water Max. 0.03%
Boiling Point 110°C to 111°C
Solubility Information 0.5g/L in water
Formula Weight 92.14g/mol
Percent Purity Min. 99.7%
Grade For Organic Residue Analysis
Assay By GC, corrected for water
Chemical Name or Material Toluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.