missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:
- Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
- Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
- Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation
Specifications
Specifications
| Melting Point | -95°C |
| CAS | 108-88-3 |
| Color | Colorless |
| Acidity | Max. 0.0005meq/g |
| Density | 0.87g/cm3 at 20°C |
| Quantity | 2.5 L |
| Explosion Limit | 1.2 - 7 Vol % |
| Flash Point | 4°C |
| Ignition Point | 535°C |
| Odor | Aromatic |
| Show More |
Safety and Handling
Signal Word
- Danger
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?