missing translation for 'onlineSavingsMsg'
Learn More

Toluene, BAKER ANALYZED, J.T. Baker™

Product Code. 10662521 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 L
Packaging:
EcoTainer Metal Safety Can
Unit Size:
5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10662521 5 L EcoTainer Metal Safety Can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10662521 Supplier JT Baker Supplier No. 8077.5000
  • ONLINE EXCLUSIVE PRICE
    £370.00
    £356.90 / 5 L
    Save £13.10
    3% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -95°C
CAS 108-88-3
Color Colorless
Density 0.87g.cm3 at 20°C
Quantity 5 L
Explosion Limit 1.2 - 7 Vol %
Flash Point 4°C
Ignition Point 535°C
Odor Aromatic
Packaging EcoTainer Metal Safety Can
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Molecular Formula C7H8
Linear Formula C6H5CH3
MDL Number MFCD00008512
Viscosity 0.6 mPas at 20°C
Water Max. 0.03%
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
Boiling Point 110°C to 111°C
Solubility Information 0.5g/L in water
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.14
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14g/mol
Percent Purity ≥99.5%
Grade Exceeds ACS specifications
Chemical Name or Material Toluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.