missing translation for 'onlineSavingsMsg'
Learn More

Toluene, BAKER ANALYZED HPLC, J.T. Baker™

Product Code. 10098810 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
1 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
Unit Size:
1 L
5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10098810 1 L Glass 1 L
10613971 5 L EcoTainer Metal Safety Can 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10098810 Supplier JT Baker Supplier No. 9351.1000
  • ONLINE EXCLUSIVE PRICE
    £194.90
    £163.26 / 1 L
    Save £31.64
    16% Off
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Get rapid, reproducible performance and separation results. Toluene, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Melting Point -95°C
Absorbance Max. 0.40 at 288nm, Max. 0.10 at 300nm, Max. 0.01 at 350nm
Color Colorless
Density 0.87/cm3
Quantity 1 L
Explosion Limit 1.2 - 7 Vol %
Flash Point 4°C
Ignition Point 535°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 3 ppm
Linear Formula C6H5CH3
Viscosity 0.6 mPas at 20°C
Water Max. 0.02%
Boiling Point 110°C to 111°C
Solubility Information 0.5g/L in water
Formula Weight 92.14g/mol
Percent Purity ≥99.7%
Grade For use in High Performance Liquid Chromatography
Chemical Name or Material Toluene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.