missing translation for 'onlineSavingsMsg'
Learn More

Toluene-4-Sulfonic Acid, Extra Pure, SLR, Fisher Chemical™

Product Code. 10589390 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10589390 100 g 100g
10686462 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10589390 Supplier Fisher Chemical Supplier No. T/2500/48
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6192-52-5
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number 142137
InChI Key KJIFKLIQANRMOU-UHFFFAOYSA-N
Synonym p-toluenesulfonic acid monohydrate, 4-methylbenzenesulfonic acid hydrate, 4-methylbenzenesulfonic acid monohydrate, p-toluene sulfonic acid monohydrate, unii-3bto78gaff, benzenesulfonic acid, 4-methyl-, monohydrate, ptoluenesulfonic acid, p-toluenesulfonic acid, monohydrate, 3bto78gaff, 4-methylbenzene-1-sulfonic acid hydrate
PubChem CID 521998
IUPAC Name 4-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC=C(C=C1)S(=O)(=O)O.O

Specifications

Melting Point 100°C
Color White
Quantity 100 g
Packaging Plastic Powder Jar
Physical Form Solid
Formula Weight 190.22g/mol
compliance-icons
Product Identifier
  • p-Toluenesulfonic acid monohydrate
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Toluene-4-sulfonic acid monohydrate, p-Toluenesulfonic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.