missing translation for 'onlineSavingsMsg'
Learn More

Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate, Thermo Scientific Chemicals

Catalog Number 11384257
Click to view available options
Quantity:
250 g
1 kg
584-84-9
C9H6N2O2
174.159
MFCD00002011
DVKJHBMWWAPEIU-UHFFFAOYSA-N
2,4-toluene diisocyanate, 2,4-diisocyanatotoluene, tolylene-2,4-diisocyanate, toluene-2,4-diisocyanate, 2,4-tolylene diisocyanate, toluene 2,4-diisocyanate, 4-methyl-m-phenylene diisocyanate, tolylene diisocyanate, 2,4-tdi, toluylene-2,4-diisocyanate
11443
CHEBI:53556
2,4-diisocyanato-1-methylbenzene
CC1=C(C=C(C=C1)N=C=O)N=C=O
584-84-9
C9H6N2O2
174.159
MFCD00002011
DVKJHBMWWAPEIU-UHFFFAOYSA-N
2,4-toluene diisocyanate, 2,4-diisocyanatotoluene, tolylene-2,4-diisocyanate, toluene-2,4-diisocyanate, 2,4-tolylene diisocyanate, toluene 2,4-diisocyanate, 4-methyl-m-phenylene diisocyanate, tolylene diisocyanate, 2,4-tdi, toluylene-2,4-diisocyanate
11443
CHEBI:53556
2,4-diisocyanato-1-methylbenzene
CC1=C(C=C(C=C1)N=C=O)N=C=O

Toluene diisocyanate is utilized in polyurethane foams and tank trucks. It is also used in concrete sealers for aircraft, sealers and coatings in floor. It is employed as a crosslinking agent for nylon.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 19.5°C to 21.5°C
Color Colorless
Density 1.225 g/mL
Boiling Point 251°C
Flash Point 132° (270°F)
Refractive Index 1.567
Quantity 250 g
UN Number UN2078
Beilstein 744602
Sensitivity Moisture sensitive
Merck Index 14,9530
Solubility Information Miscible with ether,acetone,benzene,carbontetrachloride and chlorobenzene.
Formula Weight 174.16
Percent Purity 80%
Grade Technical
Physical Form Liquid
Assay Remainder 2,6-diisocyanate
Chemical Name or Material Toluene-2,4-diisocyanate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'