missing translation for 'onlineSavingsMsg'
Learn More

Tolterodine, MedChemExpress

Product Code. 16468268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16468268 10 mg 10mg
16457996 100 mg 100mg
16454157 50 mg 50mg
16450708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16468268 Supplier Medchem Express Supplier No. HYA0024

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolterodine(PNU-200583) is a potent muscarinic receptor antagonists that show selectivity for the urinary bladder over salivary glands in vivo.

Tolterodine(PNU-200583) is a potent muscarinic receptor antagonists that show selectivity for the urinary bladder over salivary glands in vivo.

Specifications

Chemical Name or Material Tolterodine
CAS 124937-51-5
Color Colorless
Purity Grade Notes Research
Molecular Formula C22H31NO
Quantity 10 mg
Synonym (R)-(+)-Tolterodine (+)-Tolterodine (R)-Tolterodine PNU-200583
Solubility Information Ethanol : 120 mg/mL (368.67 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (307.23 mM; Need ultrasonic)
SMILES CC(C)N(C(C)C)CC[C@H](C1=CC=CC=C1)C2=C(O)C=CC(C)=C2
Molecular Weight (g/mol) 325.49
Formula Weight 325.49
Percent Purity 95.0%
Grade Research
Physical Form Oil
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.