missing translation for 'onlineSavingsMsg'
Learn More

Tolmetin sodium dihydrate, MedChemExpress

Product Code. 16448776
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448776 Supplier Medchem Express Supplier No. HYB1489
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tolmetin sodium dihydrate is an orally active and potent COX inhibitor with IC50s of 0.35 μM and 0.82 μM human COX-1 and COX-2, respectively. Tolmetin sodium dihydrate is a non-steroidal anti-inflammatory drug (NSAID).

Tolmetin sodium dihydrate is an orally active and potent COX inhibitor with IC50s of 0.35 μM and 0.82 μM human COX-1 and COX-2, respectively. Tolmetin sodium dihydrate is a non-steroidal anti-inflammatory drug (NSAID).

Specifications

Chemical Name or Material Tolmetin sodium dihydrate
CAS 64490-92-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C15H18NNaO5
Quantity 100 mg
Solubility Information H2O : ≥ 100 mg/mL (317.16 mM) ∣DMSO : 12.5 mg/mL (39.64 mM; Need ultrasonic)
SMILES O=C(O[Na])CC1=CC=C(C(C2=CC=C(C)C=C2)=O)N1C.O.O
Molecular Weight (g/mol) 315.3
Formula Weight 315.3
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.