missing translation for 'onlineSavingsMsg'
Learn More

Tollip Antibody (SB40a), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18432447 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432447 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432447 Supplier Novus Biologicals Supplier No. NBP128621AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tollip Monoclonal antibody specifically detects Tollip in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tollip
Applications Western Blot, ELISA
Classification Monoclonal
Clone SB40a
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein
Host Species Mouse
Immunogen Recombinant Tollip
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 54472
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.