missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolimidone, MedChemExpress
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
Tolimidone is a potent and selective allosteric activator of Lyn kinase with an EC50 of 63 nM.
Specifications
Specifications
| Chemical Name or Material | Tolimidone |
| CAS | 41964-07-2 |
| Color | Earth Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C11H10N2O2 |
| Quantity | 5 mg |
| Synonym | MLR-1023 |
| Solubility Information | DMSO : 150 mg/mL (741.80 mM; Need ultrasonic) |
| SMILES | O=C1N=CC(OC2=CC=CC(C)=C2)=CN1 |
| Molecular Weight (g/mol) | 202.21 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction