missing translation for 'onlineSavingsMsg'
Learn More

Tolfenpyrad, MedChemExpress

Product Code. 16417356
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16417356 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16417356 Supplier Medchem Express Supplier No. HY17516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolfenpyrad is a pesticide that was first approved in 2002 in Japan.

Tolfenpyrad is a pesticide that was first approved in 2002 in Japan.

Specifications

Chemical Name or Material Tolfenpyrad
CAS 129558-76-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H22ClN3O2
Quantity 100 mg
Solubility Information DMSO : 110 mg/mL (286.56 mM; Need ultrasonic)
SMILES O=C(C1=C(Cl)C(CC)=NN1C)NCC2=CC=C(OC3=CC=C(C)C=C3)C=C2
Molecular Weight (g/mol) 383.87
Formula Weight 383.87
Percent Purity 98.86%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.