missing translation for 'onlineSavingsMsg'
Learn More

Tolfenamic Acid, MedChemExpress

Product Code. p-200101083
Encompass_Preferred
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 16488448

Brand: Medchem Express HYB0335

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolfenamic Acid (GEA 6414) is a non-steroidal anti-inflammatory and anti-cancer agent, selectively inhibits COX-2, with an IC50 of 13.49 μM (3.53 μg/mL) in LPS-treated (COX-2) canine DH82 monocyte/macrophage cells, but shows no effect on COX-1.

Tolfenamic Acid (GEA 6414) is a non-steroidal anti-inflammatory and anti-cancer agent, selectively inhibits COX-2, with an IC50 of 13.49 μM (3.53 μg/mL) in LPS-treated (COX-2) canine DH82 monocyte/macrophage cells, but shows no effect on COX-1.

Specifications

Chemical Name or Material Tolfenamic Acid
Physical Form Powder
CAS 13710-19-5
Color White
Quantity 10 g
Purity Grade Notes Research
Molecular Formula C14H12ClNO2
For Use With (Application) COVID-19-immunoregulation
Synonym GEA 6414
Solubility Information DMSO : ≥ 100 mg/mL (382.12 mM) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)C1=CC=CC=C1NC2=C(C)C(Cl)=CC=C2
Molecular Weight (g/mol) 261.7
Formula Weight 261.7
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.