missing translation for 'onlineSavingsMsg'
Learn More

Tolfenamic acid, 99+%

Artikelnummer. 15473869
Ansicht ändern
Click to view available options
Quantity:
5 g
10 g
Packungsgröße:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Packungsgröße
15473869 10 g 10g
15463869 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Artikelnummer. 15473869 Lieferant Thermo Scientific Alfa Aesar Lieferanten-Nr. J61256.09
 Restricted Item
In Stock
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

NSAID that activates calcium-activated potassium channels | CAS: 13710-19-5 | C14H12ClNO2 | 261.705 g/mol

Tolfenamic acid is a non-steroidal anti-inflammatory agent. It interferes with synthesis of β-amyloid precursor protein, and thus Aβ peptides, by promoting degradation of an essential transcription factor. It inhibits fMLP- and A23187-induced Ca2+ influx in human PMNL with an IC50 value of approximately 20 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolfenamic acid is a non-steroidal anti-inflammatory agent. It interferes with synthesis of β-amyloid precursor protein, and thus Aβ peptides, by promoting degradation of an essential transcription factor. It inhibits fMLP- and A23187-induced Ca2+ influx in human PMNL with an IC50 value of approximately 20 µM.

Solubility
Soluble in water (slightly), acetone (∼10 mg/ml), DMSO (52 mg/ml at 25°C), methanol (∼10 mg/ml) and ethanol (50 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 13710-19-5
Molecular Formula C14H12ClNO2
Molecular Weight (g/mol) 261.705
MDL Number MFCD00133865
InChI Key YEZNLOUZAIOMLT-UHFFFAOYSA-N
Synonym tolfenamic acid, clotam, n-3-chloro-2-methylphenyl anthranilic acid, 2-3-chloro-2-methylphenyl amino benzoic acid, tolfedine, acido tolfenamico, acide tolfenamique, acidum tolfenamicum, tolfine, n-2-methyl-3-chlorophenyl anthranilic acid
PubChem CID 610479
ChEBI CHEBI:32243
IUPAC Name 2-(3-chloro-2-methylanilino)benzoic acid
SMILES CC1=C(C=CC=C1Cl)NC2=CC=CC=C2C(=O)O

Spezifikation

Color White
Quantity 10 g
UN Number UN2811
Merck Index 14,9513
Solubility Information Soluble in water (slightly),acetone (∽10mg/ml),DMSO (52mg/ml at 25°C),methanol (∽10mg/ml) and ethanol (50mg/ml).
Formula Weight 261.7
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material Tolfenamic acid
compliance-icons
Produktidentifikator
  • Tolfenamic acid
Signalwort
  • Danger
Gefahrenkategorie
  • Acute toxicity Category 3
Gefahrenhinweis
  • H301-Toxic if swallowed.
Sicherheitshinweis
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Ergänzende Informationen
  • MIXTURE LIST-Contain: Tolfenamic acid

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.