missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolclofos-methyl, MedChemExpress
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Tolclofos-methyl is a broad-spectrum aromatic hydrocarbon fungicide that is used as a see treatment for protection against soil-borne and seed borne fungal pathogens that caused seed decay and seedling blights.
Specifications
Specifications
| Chemical Name or Material | Tolclofos-methyl |
| Physical Form | Solid |
| CAS | 57018-04-9 |
| Color | Off-White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H11Cl2O3PS |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : ≥ 36 mg/mL (119.55 mM) |
| SMILES | S=P(OC)(OC1=C(Cl)C=C(C)C=C1Cl)OC |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction