missing translation for 'onlineSavingsMsg'
Learn More

Tolcapone, MedChemExpress

Product Code. 16427328
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16427328

Brand: Medchem Express HY17406

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tolcapone (Ro 40-7592) is a selective, orally active and powerful mixed (peripheral and central) COMT inhibitor with an IC50 of 773 nM in the liver. Tolcapone is also a potent inhibitor of α-syn and Aβ42 oligomerization and fibrillogenesis. Tolcapone induces oxidative stress leading to apoptosis and inhibition of tumor growth in neuroblastoma.

Tolcapone (Ro 40-7592) is a selective, orally active and powerful mixed (peripheral and central) COMT inhibitor with an IC50 of 773 nM in the liver. Tolcapone is also a potent inhibitor of α-syn and Aβ42 oligomerization and fibrillogenesis. Tolcapone induces oxidative stress leading to apoptosis and inhibition of tumor growth in neuroblastoma.

Specifications

Chemical Name or Material Tolcapone
CAS 134308-13-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H11NO5
Quantity 10 mg
Synonym Ro 40-7592
Solubility Information DMSO : 100 mg/mL (365.98 mM; Need ultrasonic)
SMILES O=C(C1=CC(O)=C(O)C([N+]([O-])=O)=C1)C2=CC=C(C)C=C2
Molecular Weight (g/mol) 273.24
Formula Weight 273.24
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.