missing translation for 'onlineSavingsMsg'
Learn More

Tolcapone, MedChemExpress

Product Code. 16487286
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487286 100 mg 100mg
16473337 50 mg 50mg
16427328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16487286 Supplier Medchem Express Supplier No. HY17406
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tolcapone (Ro 40-7592) is a selective, orally active and powerful mixed (peripheral and central) COMT inhibitor with an IC50 of 773 nM in the liver. Tolcapone is also a potent inhibitor of α-syn and Aβ42 oligomerization and fibrillogenesis. Tolcapone induces oxidative stress leading to apoptosis and inhibition of tumor growth in neuroblastoma.

Tolcapone (Ro 40-7592) is a selective, orally active and powerful mixed (peripheral and central) COMT inhibitor with an IC50 of 773 nM in the liver. Tolcapone is also a potent inhibitor of α-syn and Aβ42 oligomerization and fibrillogenesis. Tolcapone induces oxidative stress leading to apoptosis and inhibition of tumor growth in neuroblastoma.

Specifications

Chemical Name or Material Tolcapone
CAS 134308-13-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C14H11NO5
Quantity 100 mg
Synonym Ro 40-7592
Solubility Information DMSO : 100 mg/mL (365.98 mM; Need ultrasonic)
SMILES O=C(C1=CC(O)=C(O)C([N+]([O-])=O)=C1)C2=CC=C(C)C=C2
Molecular Weight (g/mol) 273.24
Formula Weight 273.24
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.