missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolcapone-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Tolcapone-d4 |
| CAS | 1246816-93-2 |
| Molecular Formula | C14H7D4NO5 |
| Quantity | 1 mg |
| Synonym | (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl-d4)methanone, Ro-40-7592-d4, Tasmar-d4 |
| InChI Formula | InChI=1S/C14H11NO5/c1-8-2-4-9(5-3-8)13(17)10-6-11(15(19)20)14(18)12(16)7-10/h2-7,16,18H,1H3/i2D,3D,4D,5D |
| SMILES | O=C(C1=C([2H])C([2H])=C(C)C([2H])=C1[2H])C2=CC(O)=C(O)C([N+]([O-])=O)=C2 |
| Molecular Weight (g/mol) | 277.27 |
| Formula Weight | 277.0888 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction