missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Tolcapone
GREENER_CHOICE

Product Code. 18713188
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18713188

Brand: Tocris Bioscience™ 5864/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

COMT inhibitor; also inhibits transthyretin aggregation

Tolcapone is a COMT inhibitor. Inhibits both brain and peripheral COMT. Also binds transthyretin (TTR) with high affinity (Kd1 and Kd2 values are 21 and 58 nM, respectively). Inhibits TTR aggregation in human plasma and prevents TTR-induced cytotoxicity in vitro. Stabilizes TTR in mice and humans in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3,4-Dihydroxy-5-nitrophenyl)(4-methylphenyl)methanone
CAS 134308-13-7
Synonym Tasmar, Ro 40-7592
Quantity 50 mg
Target Catechol O-Methyltransferase (COMT) Inhibitors
Molecular Formula C14H11NO5
Formula Weight 273.24
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.