missing translation for 'onlineSavingsMsg'
Learn More

Tolbutamide, Tolbutamide, TRC

Product Code. 30136526
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136526 1 g 1g
30009116 500 mg 500mg
17932115 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136526 Supplier Toronto Research Chemicals Supplier No. TRCT5351501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 H18 N2 O3 S
Molecular Weight (g/mol) 270.35
Synonym Tolbutamide, Benzenesulfonamide, N-[(butylamino)carbonyl]-4-methyl-, Urea, 1-butyl-3-(p-tolylsulfonyl)- (8CI), N-[(Butylamino)carbonyl]-4-methylbenzenesulfonamide, 1-Butyl-3-(4-methylphenyl)sulfonylurea, 1-Butyl-3-(p-methylphenylsulfonyl)urea, 1-Butyl-3-(p-tolylsulfonyl)urea, 3-(p-Tolyl-4-sulfonyl)-1-butylurea, 3-Butyl-1-(4-methylbenzenesulfonyl)urea, Aglicid, Arkozal, Artosin, Artozin, Butamid, Butamide, D 860, Diaben, Diabetamid, Diabetol, Diabuton, Diasulfon, Dolipol, Glyconon, HLS 831, Ipoglicone, Mobenol, N-(4-Methylbenzenesulfonyl)-N'-butylurea, N-(4-Methylphenylsulfonyl)-N'-butylurea, N-(Sulfonyl-p-methylbenzene)-N'-n-butylurea, N-(p-Methylbenzenesulfonyl)-N'-butylurea, N-(p-Tolylsulfonyl)-N'-butylcarbamide, N-Butyl-N'-(4-methylphenylsulfonyl)urea, N-Butyl-N'-(p-tolylsulfonyl)urea, N-Butyl-N'-p-toluenesulfonylurea, N-n-Butyl-N'-tosylurea, NSC 23813, NSC 87833, Orabet, Oralin, Orezan, Orinase, Orinaz, Oterben, Pramidex, Rastinon, Tolbusal, Tolbutamid, Tolbutamide, Toluina, Tolumid, Tolumide, Toluvan, U 2043, Willbutamide
IUPAC Name 1-butyl-3-(4-methylphenyl)sulfonylurea
SMILES CCCCNC(=O)NS(=O)(=O)c1ccc(C)cc1

Specifications

CAS 64-77-7
Quantity 1 g
InChI Formula InChI=1S/C12H18N2O3S/c1-3-4-9-13-12(15)14-18(16,17)11-7-5-10(2)6-8-11/h5-8H,3-4,9H2,1-2H3,(H2,13,14,15)
Formula Weight 270.1038
Percent Purity >95%
Chemical Name or Material Tolbutamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.