missing translation for 'onlineSavingsMsg'
Learn More

Tolbutamide, 98%

Product Code. 11480287
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480287 250 g 250g
11460287 10 g 10g
11470287 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480287 Supplier Thermo Scientific Alfa Aesar Supplier No. B21698.30
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64-77-7 | C12H18N2O3S | 270.347 g/mol

Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tolbutamide, have been used in a cDNA microarray assay to probe changes in gene expression in HepG2 cells upon their administration. It has been utilized to counteract insulin activity in a patch-clamp investigation of ATP sensitive K+ channels in mouse pancreatic β-cells. The activity of various biotransformation enzymes in cultured primary rat proximal tubular cells in the presence of tolbutamide and other compounds has been studied.

Solubility
Soluble in chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-77-7
Molecular Formula C12H18N2O3S
Molecular Weight (g/mol) 270.347
MDL Number MFCD00027169
InChI Key JLRGJRBPOGGCBT-UHFFFAOYSA-N
Synonym tolbutamide, orinase, 1-butyl-3-tosylurea, tolbutamid, aglicid, arkozal, artosin, diabetol, dolipol, rastinon
PubChem CID 5505
ChEBI CHEBI:27999
IUPAC Name 1-butyl-3-(4-methylphenyl)sulfonylurea
SMILES CCCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)C

Specifications

Melting Point 128°C to 130°C
Quantity 250 g
Beilstein 1984428
Merck Index 14,9507
Solubility Information Soluble in chloroform.
Formula Weight 270.35
Percent Purity 98%
Chemical Name or Material Tolbutamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.