missing translation for 'onlineSavingsMsg'
Learn More

Tolazoline, TRC

Product Code. 30451459
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451459 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30451459 Supplier Toronto Research Chemicals Supplier No. TRCT53500310G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tolazoline
CAS 59-98-3
Molecular Formula C10H12N2
Quantity 10 g
Synonym 2-Imidazoline','2-benzyl- (8CI), Imidazoline','2-benzyl- (6CI), 4,5-Dihydro-2-(phenylmethyl)-1H-imidazole, 2-Benzyl-2-imidazoline, 2-Benzyl-4,5-dihydro-1H-imidazole, 2-Benzyl-4,5-imidazoline, 2-Benzylimidazoline, Artonil, Benzazoline, Benzidazol, Benzylimidazoline, Dilatol ASI, Divascol, Imidalin, Imidaline, Kasimid, Lambril, NSC 35110, Olitensol, Peripherine, Phenylmethylimidazoline, Prefaxil, Priscol, Priscoline, Vasimid, Vasodil
InChI Formula InChI=1S/C10H12N2/c1-2-4-9(5-3-1)8-10-11-6-7-12-10/h1-5H,6-8H2,(H,11,12)
SMILES C(C1=NCCN1)c2ccccc2
IUPAC Name 2-benzyl-4,5-dihydro-1H-imidazole
Molecular Weight (g/mol) 160.22
Formula Weight 160.1
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.