missing translation for 'onlineSavingsMsg'
Learn More

Tolazoline hydrochloride, MedChemExpress

Product Code. p-200099044
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16470718

Brand: Medchem Express HYA0066A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Tolazoline (hydrochloride)(Imidaline (hydrochloride)) Hcl is a non-selective competitive α-adrenergic receptor antagonist.

Tolazoline (hydrochloride)(Imidaline (hydrochloride)) Hcl is a non-selective competitive α-adrenergic receptor antagonist.

Specifications

Chemical Name or Material Tolazoline hydrochloride
Physical Form Solid
CAS 59-97-2
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H13ClN2
Synonym Imidaline hydrochlorideNSC35110 hydrochloride
Solubility Information H2O : ≥ 50 mg/mL (254.22 mM) ∣DMSO : 33.33 mg/mL (169.46 mM; Need ultrasonic)
SMILES [H]Cl.C1(CC2=CC=CC=C2)=NCCN1
Molecular Weight (g/mol) 196.68
Formula Weight 196.68
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.