missing translation for 'onlineSavingsMsg'
Learn More

Tolazamide, TRC

Product Code. 30457106
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457106 250 mg 250mg
30449122 100 mg -
30449917 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457106 Supplier Toronto Research Chemicals Supplier No. TRCT534500250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1156-19-0
Molecular Formula C14 H21 N3 O3 S
Molecular Weight (g/mol) 311.4
Synonym Benzenesulfonamide, N-[[(hexahydro-1H-azepin-1-yl)amino]carbonyl]-4-methyl-, Urea, 1-(hexahydro-1H-azepin-1-yl)-3-(p-tolylsulfonyl)- (6CI,7CI,8CI), 1-(4-Methylphenylsulfonyl)-3-(hexahydro-1H-azepin-1-yl)urea, 1-(Hexahydro-1-azepinyl)-3-p-tolylsulfonylurea, 1-(Hexahydro-1H-azepin-1-yl)-3-(p-tolylsulfonyl)urea, Diabewas, N-(p-Toluenesulfonyl)-N'-hexamethyleniminourea, NSC 70762, Norglycin, Tolanase, Tolazamide, Tolinase, U 17835
IUPAC Name 1-(azepan-1-yl)-3-(4-methylphenyl)sulfonylurea
SMILES Cc1ccc(cc1)S(=O)(=O)NC(=O)NN2CCCCCC2

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C14H21N3O3S/c1-12-6-8-13(9-7-12)21(19,20)16-14(18)15-17-10-4-2-3-5-11-17/h6-9H,2-5,10-11H2,1H3,(H2,15,16,18)
Formula Weight 311.1304
Percent Purity >95%
Chemical Name or Material Tolazamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.