missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolazamide-d7, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Tolazamide-d7 |
| CAS | 1794811-72-5 |
| Molecular Formula | C14H14D7N3O3S |
| Quantity | 2.5 mg |
| Synonym | N-[[(Hexahydro-1H-azepin-1-yl)amino]carbonyl]-4-methylbenzenesulfonamide-d7, 1-(4-Methylphenyl-d7-sulfonyl)-3-(hexahydro-1H-azepin-1-yl)urea, 1-(Hexahydro- 1-azepinyl)-3-p-tolylsulfonylurea-d7, Diabewas-d7, N-(p-Toluenesulfonyl)-N'-hexa- methyleniminourea-d7, NSC 70762, -d7 Norglycin-d7, Tolanase-d7, Tolinase-d7, U 17835-d7 |
| InChI Formula | InChI=1S/C14H21N3O3S/c1-12-6-8-13(9-7-12)21(19,20)16-14(18)15-17-10-4-2-3-5-11-17/h6-9H,2-5,10-11H2,1H3,(H2,15,16,18)/i1D3,6D,7D,8D,9D |
| SMILES | O=S(C1=C([2H])C([2H])=C(C([2H])([2H])[2H])C([2H])=C1[2H])(NC(NN2CCCCCC2)=O)=O |
| Molecular Weight (g/mol) | 318.44 |
| Formula Weight | 318.1743 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction