missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TokeOni

Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
TokeOni is a near-infrared red (NIR)-emission luciferin analog. Peak luminescence at 677 nM. ∼40-fold brighter than the luciferin-luciferase system in vitro, and 100-1000 times brighter in vivo. Allows non-invasive visualization of single cells deep inside freely moving animals. Ideal for deep tissue applications. Water soluble (<40 mM), orally bioavailable and brain penetrant.
This product is supplied as a hydrochloride salt, the stoichiometry of which is batch dependant. Please refer to the Certificate of Analysis to obtain the batch specific Net Product Content.
Specifications
Specifications
| Chemical Name or Material | (4S)-2-[(1E,3E)-4-[4-(Dimethylamino)phenyl]-1,3-butadien-1-yl]-4,5-dihydro-4-thiazolecarboxylic acid hydrochloride |
| Synonym | AkaLumine HCl |
| Quantity | 5 mg |
| Target | Bioluminescent Substrates |
| Molecular Formula | C16H18N2O2S.xHCl |
| Formula Weight | 338.85 |
| Purity | 90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction