missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BCCIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565200
This item is not returnable.
View return policy
Description
Immunogen sequence: ALPMYQQLQKE LAGAHRTNKP CGKCYFYLLI SKTFVEAGKN NSKKKPSNKK KAALMFANAE EEFFYEEQGK PEVLGGPDTR RGLEPVPIQH NGGSR Highest antigen sequence identity to the following orthologs - mouse 57%, rat 54%.
This gene product was isolated on the basis of its interaction with BRCA2 and p21 proteins. It is an evolutionarily conserved nuclear protein with multiple interacting domains. The N-terminal half shares moderate homology with regions of calmodulin and M-calpain, suggesting that it may also bind calcium. Functional studies indicate that this protein may be an important cofactor for BRCA2 in tumor suppression, and a modulator of CDK2 kinase activity via p21. This protein has also been implicated in the regulation of BRCA2 and RAD51 nuclear focus formation, double-strand break-induced homologous recombination, and cell cycle progression. Multiple transcript variants encoding different isoforms have been described for this gene.
Specifications
| BCCIP | |
| Polyclonal | |
| Unconjugated | |
| BCCIP | |
| 1110013J05Rik; Bccip; BCCIPalpha; BCCIPbeta; BRCA2 and CDKN1A interacting protein; BRCA2 and CDKN1A-interacting protein; BRCA2 and Cip1/p21 interacting protein; cdk inhibitor p21 binding protein; p21- and CDK-associated protein 1; Protein TOK-1; TOK1; TOK-1; TOK-1alpha; TOK-1beta | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56647 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P287 | |
| BCCIP | |
| Recombinant Human TOK-1. Recombinant protein control fragment (Product #RP-105943). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction