missing translation for 'onlineSavingsMsg'
Learn More

Tofogliflozin, TRC

Product Code. 30453411
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30453411 25 mg 25mg
30453853 100 mg 100mg
30452100 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30453411 Supplier Toronto Research Chemicals Supplier No. TRCT52875025MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903565-83-3
Molecular Formula C22 H26 O6
Molecular Weight (g/mol) 386.44
Synonym (1S,3'R,4'S,5'S,6'R)-6-[(4-Ethylphenyl)methyl]-3',4',5',6'-tetrahydro-6'-(hydroxymethyl)spiro[isobenzofuran-1(3H),2'-[2H]pyran]-3',4',5'-triol, CSG 452
IUPAC Name (3'R,4'S,5'S,6'R)-5-[(4-ethylphenyl)methyl]-6'-(hydroxymethyl)spiro[1H-2-benzofuran-3,2'-oxane]-3',4',5'-triol
SMILES CCc1ccc(Cc2ccc3COC4(O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)c3c2)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C22H26O6/c1-2-13-3-5-14(6-4-13)9-15-7-8-16-12-27-22(17(16)10-15)21(26)20(25)19(24)18(11-23)28-22/h3-8,10,18-21,23-26H,2,9,11-12H2,1H3/t18-,19-,20+,21-,22?/m1/s1
Formula Weight 386.1729
Chemical Name or Material Tofogliflozin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.