missing translation for 'onlineSavingsMsg'
Learn More

Tofenacin Hydrochloride Salt, TRC

Product Code. 30456562
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30456562 5 mg 5mg
30452484 10 mg -
30451299 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456562 Supplier Toronto Research Chemicals Supplier No. TRCT5285005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10488-36-5
Molecular Formula C17 H21 N O . Cl H
Molecular Weight (g/mol) 291.82
Synonym N-Desmethylorphenadrine Hydrochloride
IUPAC Name N-methyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
SMILES Cl.CNCCOC(c1ccccc1)c2ccccc2C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C17H21NO.ClH/c1-14-8-6-7-11-16(14)17(19-13-12-18-2)15-9-4-3-5-10-15;/h3-11,17-18H,12-13H2,1-2H3;1H
Formula Weight 291.139
Chemical Name or Material N-Desmethylorphenadrine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.