missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tofacitinib, MedChemExpress
Description
Tofacitinib is an orally available JAK3/2/1 inhibitor with IC50s of 1, 20, and 112 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Tofacitinib |
| CAS | 477600-75-2 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C16H20N6O |
| Quantity | 500 mg |
| Synonym | Tasocitinib CP-690550 |
| Solubility Information | DMSO : 125 mg/mL (400.17 mM; Need ultrasonic) ∣Ethanol : 2.5 mg/mL (8.00 mM; Need ultrasonic) ∣H2O : 0.15 mg/mL (0.48 mM; Need ultrasonic and warming) |
| SMILES | O=C(CC#N)N1C[C@H](N(C2=C3C(NC=C3)=NC=N2)C)[C@H](C)CC1 |
| Molecular Weight (g/mol) | 312.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?