missing translation for 'onlineSavingsMsg'
Learn More

Tofacitinib impurity N, TRC

Product Code. 30455854
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455854 5 mg 5mg
30455510 10 mg 10mg
30451693 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30455854 Supplier Toronto Research Chemicals Supplier No. TRCT5280205MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1260614-73-0
Molecular Formula C13H19N5
Molecular Weight (g/mol) 245.32
Synonym N-Methyl-N-((3S,4S)-4-methylpiperidin-3-yl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine
IUPAC Name N-methyl-N-[(3S,4S)-4-methylpiperidin-3-yl]-7H-pyrrolo[2,3-d]pyrimidin-4-amine
SMILES C[C@H]1CCNC[C@H]1N(C)c2ncnc3[nH]ccc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C13H19N5/c1-9-3-5-14-7-11(9)18(2)13-10-4-6-15-12(10)16-8-17-13/h4,6,8-9,11,14H,3,5,7H2,1-2H3,(H,15,16,17)/t9-,11+/m0/s1
Formula Weight 245.164
Chemical Name or Material Tofacitinib impurity N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.