missing translation for 'onlineSavingsMsg'
Learn More

Tofacitinib citrate, MedChemExpress

Product Code. 16491306
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16491306

Brand: Medchem Express HY40354A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tofacitinib citrate is an orally available JAK1/2/3 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Tofacitinib citrate has antibacterial, antifungal and antiviral activities.

Tofacitinib citrate is an orally available JAK1/2/3 inhibitor with IC50s of 1, 20, and 112 nM, respectively. Tofacitinib citrate has antibacterial, antifungal and antiviral activities.

Specifications

Chemical Name or Material Tofacitinib citrate
Physical Form Solid
CAS 540737-29-9
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C22H28N6O8
For Use With (Application) COVID-19-anti-virus
Synonym Tasocitinib citrateCP-690550 citrate
Solubility Information DMSO : 28.57 mg/mL (56.63 mM; Need ultrasonic) ∣H2O : 3.33 mg/mL (6.60 mM; Need ultrasonic)
SMILES O=C(CC#N)N1C[C@H](N(C2=C3C(NC=C3)=NC=N2)C)[C@H](C)CC1.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 504.49
Formula Weight 504.49
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.