missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TOE1 Polyclonal antibody specifically detects TOE1 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TOE1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | FLJ13949, target of EGR1 protein 1, target of EGR1, member 1 (nuclear) |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 460 and 510 of human Target of EGR1, Member 1 using the numbering given in entry NP_079353.3 (GeneID 114034). |
| Purification Method | Immunogen affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?