Learn More
Tocris Bioscience™ ProTx II
GREENER_CHOICE
Product Code. 18055222
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. 18055222
Brand: Tocris Bioscience™ 4023/100U
Selective NaV1.7 channel blocker
ProTx II is a selective NaV1.7 channel blocker. Shifts activation gating positively and decreases current magnitude. Displays 100-fold selectivity over other sodium channel subtypes.