missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Kisspeptin 10 (human)

Description
Kisspeptin 10 (human) is a potent endogenous ligand for the Kisspeptin receptor (KISS1, GPR54). Binds with high affinity to rat and human KISS1 receptors with Ki values of 1.59 and 2.33 nM respectively. Inhibits metastasis and invasion in mouse melanomas and stimulates gonadotropin secretion following i.c.v. administration.
Analog also available.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?