missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ JMV 449

Description
JMV 449 is a potent, metabolically stable neurotensin receptor agonist peptide (IC50 = 0.15 nM for inhibition of [125I]-NT binding to neonatal mouse brain; EC50 = 1.9 nM for contraction of guinea pig ileum). Produces long-lasting hypothermic, neuroprotective and analgesic effects in mice following central administration in vivo.
Specifications
Specifications
| Purity | 0.95 |
| Content And Storage | Store at -20°C |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?